Anti-NMDAR2B Rabbit Polyclonal Antibody
Catalog # 10209-474
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor Ionotropic N-methyl D-aspartate 2B
- Antigen Symbol:NMDAR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:hNR3|MGC142178|GRIN2B|NR3|MGC142180|NR2B|Glutamate Receptor ionotropic N-methyl D-aspartate 2B|NMDAR2B
- Amino Acid Number:1307 to 1320
- Sequence:FVDLQKEEAALAPR
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human NMDAR2B (1307-1320aa FVDLQKEEAALAPR), identical to the related mouse and rat sequences.
- Purification:purified by immunogen affinity purification
- Cat. No.:10209-474
- Supplier no.:PA1059-1
Specifications
About this item
Rabbit IgG polyclonal antibody for Glutamate receptor ionotropic, NMDA 2B(GRIN2B) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: NMDAR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat