Anti-NFkB p105/p50 Rabbit Polyclonal Antibody
10209-216
- Antibody Type:Primary
- Antigen Name:Nuclear factor NF-kappa-B p105 subunit
- Antigen Symbol:NFkB p105/p50
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:EBP-1|Nuclear factor NF-kappa-B p50 subunit|Nuclear factor NF-kappa-B p105 subunit|NFKB1|DNA-binding factor KBF1|Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
- Amino Acid Number:193 to 208
- Sequence:KELIRQAALQQTKEMD
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human NFkB p105 (193-208aa KELIRQAALQQTKEMD), different from the related rat sequence by one amino acid and from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. No.:10209-216
- Supplier no.:PA2111-1
Rabbit IgG polyclonal antibody for Nuclear factor NF-kappa-B p105 subunit(NFKB1) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: NFkB p105/p50
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat