Anti-Milk Fat Globule 1 Rabbit Polyclonal Antibody
Catalog # 10209-110
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:Milk Fat Globule 1
- Antigen Symbol:Milk Fat Globule 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:Lyophilized
- Amino Acid Number:165 to 180
- Sequence:NGHEFDFIHDVNKKHK
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Human Milk Fat Globule 1 (165-180aa NGHEFDFIHDVNKKHK).
- Purification:purified by immunogen affinity purification
- Cat. No.:10209-110
- Supplier no.:PA1945
Specifications
About this item
Rabbit IgG polyclonal antibody for Lactadherin(MFGE8) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Milk Fat Globule 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human