Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-FSTL3 Rabbit Polyclonal Antibody
Anti-FSTL3 Rabbit Polyclonal Antibody
Catalog # 10206-940
undefined
Anti-FSTL3 Rabbit Polyclonal Antibody
Catalog # 10206-940
Supplier Number:  PA1915

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Follistatin-related Protein 3
  • Antigen Symbol:
    FSTL3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Antigen Synonyms:
    FSRP|FLRG
  • Amino Acid Number:
    145 to 159
  • Sequence:
    ECELRAARCRGHPDL
  • Storage Temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human FSTL3 (145-159aa ECELRAARCRGHPDL), different from the related rat and mouse sequences by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Cat. No.:
    10206-940
  • Supplier no.:
    PA1915

Specifications

About this item

Rabbit IgG polyclonal antibody for Follistatin-related protein 3(FSTL3) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: FSTL3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat