Anti-NMDAR1 Rabbit Polyclonal Antibody
Catalog # 10206-684
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor ionotropic N-methyl D-aspartate 1
- Antigen Symbol:NMDAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:NMDAR1|NMD-R1|MRD8|GluN1|glutamate [NMDA] receptor subunit zeta 1|NMDA 1|subunit zeta-1|glutamate receptor ionotropic|RP11-350O14.1|NR1|glutamate [NMDA] receptor subunit zeta-1|NMDA1|N-methyl-D-aspartate receptor subunit NR1|N-methyl-D-aspartate receptor channel
- Amino Acid Number:36 to 53
- Sequence:RKHEQMFREAVNQANKRH
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human NMDAR1 (36-53aa RKHEQMFREAVNQANKRH), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. No.:10206-684
- Supplier no.:PA1222
Specifications
About this item
Rabbit IgG polyclonal antibody for Glutamate receptor ionotropic, NMDA 1(GRIN1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: NMDAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat