Anti-AFP Mouse Monoclonal Antibody [clone: AP-1]
Catalog # 10206-186
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:Alpha 1 Fetoprotein Receptor
- Antigen Symbol:AFP
- Clonality:Monoclonal
- Clone:AP-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen Synonyms:FETA|Alpha-1-Fetoprotein|HPAFP
- Storage Temperature:–20 °C
- Immunogen:Human alpha-fetoprotein.
- Cat. No.:10206-186
- Supplier no.:MA1001
Specifications
About this item
Mouse IgG monoclonal antibody for AFP, alpha-fetoprotein (AFP) detection. Tested with IHC-P in Human. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: AP-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human