Order Entry
United States
ContactUsLinkComponent
Anti-MAP2 Mouse Monoclonal Antibody [clone: MP-2]
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Microtubule-associated protein 2
  • Antigen Symbol:
    MAP2
  • Clonality:
    Monoclonal
  • Clone:
    MP-2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Antigen Synonyms:
    MAP2A|MAP2C|MAP2B
  • Storage Temperature:
    –20 °C
  • Immunogen:
    Rat brain microtubule-associated proteins (MAPs).
  • Cat. No.:
    10206-164
  • Supplier no.:
    MA1057

Specifications

About this item

Mouse IgG monoclonal antibody for MAP2, microtubule-associated protein 2 (MAP2) detection. Tested with WB, IHC-P in Human;mouse;rat. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: MP-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse