Anti-c-Myc Mouse Monoclonal Antibody [clone: IMD-3]
Catalog # 10206-122
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Symbol:c-Myc
- Clonality:Monoclonal
- Clone:IMD-3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage Temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to residues 408-439 of the human p62c-Myc protein.
- Cat. No.:10206-122
- Supplier no.:MA1028
Specifications
About this item
Mouse IgG monoclonal antibody for c-Myc, v-myc myelocytomatosis viral oncogene homolog (avian) (MYC) detection. Tested with WB, IHC-P, ICC in Human. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: IMD-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: