Anti-DLD Rabbit Polyclonal Antibody
10205-984
- Antibody Type:Primary
- Antigen Name:Dihydrolipoamide Dehydrogenase
- Antigen Symbol:DLD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:DLDH|GCSL|DLDD|Lipoamide Dehydrogenase [Porcine Heart]|LAD|Dihydrolipoyl dehydrogenase mitochondrial|E3|Dihydrolipoamide Dehydrogenase|PHE3
- Amino Acid Number:492 to 509
- Sequence:EAFREANLAASFGKSINF
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Lipoamide Dehydrogenase (492-509aa EAFREANLAASFGKSINF), different from the related mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. No.:10205-984
- Supplier no.:PA1463
Rabbit IgG polyclonal antibody for Dihydrolipoyl dehydrogenase, mitochondrial(DLD) detection. Tested with WB, IHC-P; IHC-F; ICC in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: DLD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat