Upchurch Scientific® Inlet Solvent Filters, IDEX Health & Science
21511-004
- Description:10 µm Inlet solvent filter with stem for general use, For 1/16" ID tubing
- Cat. No.:21511-004
- Supplier no.:A-302
The filters with stems are 100% 316 stainless steel.
- Filters are disposable, easy to replace without tools
- Max. flow rate 40 ml/min