EnteroScreen 4™, System for Salmonella and Shigella Screening, Hardy Diagnostics
Catalog # 10324-472
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:EnteroScreen 4™ for SS
- Cat. No.:10324-472
- Supplier no.:L225
Specifications
About this item
For the screening of enteric pathogens and the differentiation of Salmonella and Shigella from stool specimens and to rule out normal enteric flora
With this method, urease activity, lysine-decarboxylase, lysine-deaminase, gas production, and hydrogen sulfide production reactions can be simultaneously evaluated; all from one tube! Invented by Dr. Gary Procop of the Cleveland Clinic. See technical insert for color ID chart.