Order Entry
Puerto Rico
ContactUsLinkComponent
Barium iodide, ultra dry ≥99.999% (metals basis), beads -10 Mesh
Irritant
Barium iodide, ultra dry ≥99.999% (metals basis), beads -10 Mesh
Catalog # AA47118-14
CAS Number:  
Irritant
Barium iodide, ultra dry ≥99.999% (metals basis), beads -10 Mesh
Catalog # AA47118-14
Supplier Number:  047118-14
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Ampoule
  • Size:
    25 g
  • Cat. No.:
    AA47118-14
  • Supplier no.:
    47118-14

Specifications

Specification Test Results

  • Assay Minimum:
    99.999

Specification Test Results

About this item

 :
Formula:BaI₂
MDL Number:MFCD00003452
 :391.14
Merck Index:14,980
Melting Pt:740 °C
Density:5.15
Storage Temperature:Ambient
EINECS:237-276-9
UN:1564
ADR:6.1,III
 :ampouled under argon
 :GHS07
CAS Number:13718-50-8