Mouse DMP-1 PicoKine ELISA Kit, Boster
Catalog # 10205-682
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Size:96 well
- Immunogen:Standard: NSO, L17-Y503
- Cat. No.:10205-682
- Supplier no.:EK1290
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Mouse DMP-1
Ordering information: Kits include 96-well plate precoated with anti- mouse DMP-1 antibody 1; ST0000-10 Lyophilized mouse DMP-1 standard 10ng/tubex2; AR1107 Biotinylated anti- mouse DMP-1 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex (ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.