Rat sL-Selectin PicoKine ELISA Kit, Boster
Catalog # 10205-496
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Rat
- Target protein:sL-selectin
- Size:96 well
- Immunogen:Standard: NSO, W39-N332
- Cat. No.:10205-496
- Supplier no.:EK0937
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Rat sL-Selectin
Ordering information: Kits include 96-well plate precoated with anti- rat sL-Selectin antibody 1; ST0000-10 Lyophilized recombinant rat sL-Selectin standard 40ng/tubex2; AR1107 Biotinylated anti- rat sL-Selectin antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.