Human TNFsR I PicoKine ELISA Kit, Boster
Catalog # 10209-010
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Size:96 well
- Immunogen:Standard: NSO,L30-T211
- Cat. No.:10209-010
- Supplier no.:EK0528
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Human TNFsR I
Ordering information: Kits include 96-well plate precoated with anti- human TNFsR I antibody 1; ST0000-10 Lyophilized recombinant human TNFsR I standard 10ng/tubex2; AR1107 Biotinylated anti- human TNFsR I antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.