VERIFY™ Bowie-Dick Test Pack, STERIS®
Supplier: Steris
Certificates
About this item
Bowie-Dick Tests verify that the prevacuum sterilizer effectively removes air and provides efficient steam penetration.
- Reliable performance and accurate results
- Ink is free of lead and other heavy metals
- Operates at 270 - 275 °F/132 - 135 °C with an exposure time of 3.5 - 4.0 minutes
The VERIFY™ Bowie-Dick Test Pack consists of a series of air removal and steam penetration barriers into which a chemical indicator sheet is embedded. The test pack is placed directly into an otherwise empty steam sterilizer chamber and does not require a retaining device. During processing, steam must displace the air through the barrier material within the pack. A uniform change from yellow to blue on the indicator sheet indicates that all the air was removed and replaced by steam. The advanced technology in thermochromic ink formulation can detect problems with steam quality.
Certifications: Meets ANSI/AAMI/ISO 11140-4 requirements for Class 2 Air Removal Indicator Systems.