- Gradient:-
- Description:QTower Quantitative Real-Time rapidPCR Thermal Cycler with Free Color Module 1 and Laptop
- Cooling Rate:8°C/sec max, (0.1 to 8°C/sec)
- Control Accuracy:±0.2 °C
- W×D×H:240×255×430 mm
- Lid Temperature Range:Heated up to 120 °C
- Sample capacity:5-20 μl
- Cat. No.:10222-322
- Electrical:100/240 V
- Supplier no.:CPQ-8818
- Details:Ultrathin-walled 96 well microplate in parallel (low profile)
2015 Promotion Offer–Purchase a qTOWER thermal cycler and receive a free Color Module 1 and Laptop
qTOWER qPCR for quantitative Real- Time rapidPCR with fluorescence detection via 8 optical light fibers. System features gold-plated silver block and fiber optics which achieve high signal intensities and data acquisition. Heat/cool rate: 12°C/second & 8°C/second. Block temp: 4°-105°C, lid heats to 120°C. No Gradient. Color Module 1, for dyes such as FAMTM, Sybr®Green, Alexa488®, included with special offer. Select from other optional color and FRET excitation modules to meet specific wavelength requirements. Modules are placed in up to 4 positions.
The significant reduction of sample volumes (down to 5μl) provides exceptional savings of up to 75% of expensive real-time reagents. Reactions volumes of 5 - 20μl.
System features qPCRsoft quantitative analysis software (MIQE compliant) which is loaded onto the free laptop included with this promotion. The program can determine PCR efficiencies and perform absolute and relative quantifications as well as delta-delta Ct method and allele discrimination.
10 years on optics components; 2 years on the system and thermal block.
Offer runs 1/1/15 - 12/31/15. qTOWER has a small footprint and is designed for rapid qPCR of 96 samples in less than 25 minutes. Read out is in just 4 seconds, regardless of the number of colors measured.