Salmonella Minnesota Lipid A
Catalog # 89152-792
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Species:Salmonella minnesota
- Size:2 ml
- Enzyme Name:Lipid A
- Purity:≥99.9% (regarding protein or DNA contaminants; KDO not detectable)
- Concentration:1 mg/ml
- Cat. No.:89152-792
- Supplier no.:581-201-L002
Specifications
About this item
Isolated from Salmonella minnesota R595 (Re mutant)
Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water.
Prepared by mild acid hydrolysis from purified Salmonella minnesota R595 (Re) LPS (isolated by a modification of the PCP extraction method), converted to the uniform salt form and dissolved in sterile pyrogen-free double distilled water.