Order Entry
Puerto Rico
ContactUsLinkComponent
EpiXtract® Nuclear Protein Isolation Kit, Enzo Life Sciences
EpiXtract® Nuclear Protein Isolation Kit, Enzo Life Sciences
Catalog # 10135-966
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
EpiXtract® Nuclear Protein Isolation Kit, Enzo Life Sciences
Catalog # 10135-966
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-45016-0100
CAS Number:  

Specifications

  • Description:
    EpiXtract® Nuclear Protein Isolation Kit
  • Cat. No.:
    10135-966
  • No. of reactions:
    100
  • Supplier no.:
    45016-0100

Specifications

About this item

Rapid, reliable method to isolate nuclear proteins in mammalian cells and tissues.

For the extraction of nuclear proteins from mammalian cells or tissues.

EpiXtract® Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nuclear proteins.

Ordering information: Contains Pre-extraction Buffer, Extraction Buffer, Protease Inhibitor Cocktail, DTT.