Order Entry
Puerto Rico
ContactUsLinkComponent
EpiXtract® Total Histone Extraction Kit, Enzo Life Sciences
EpiXtract® Total Histone Extraction Kit, Enzo Life Sciences
Catalog # 10135-962
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
EpiXtract® Total Histone Extraction Kit, Enzo Life Sciences
Catalog # 10135-962
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-45014-0100
CAS Number:  

Specifications

  • Description:
    EpiXtract® Total Histone Extraction Kit
  • Cat. No.:
    10135-962
  • No. of reactions:
    100
  • Supplier no.:
    45014-0100

Specifications

About this item

Rapid, reliable method to isolate histone proteins in mammalian cells and tissues for histone modification applications.

For the extraction of total histone proteins from mammalian cells or tissues.

EpiXtract® Total Histone Extraction Kit provides a simple and reliable method to isolate histone proteins in 1 hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.

Ordering information: Contains 10x Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .