Order Entry
Puerto Rico
ContactUsLinkComponent
Organelle-ID RGB® Reagent IV for Microscopy, Enzo Life Sciences
Organelle-ID RGB® Reagent IV for Microscopy, Enzo Life Sciences
Catalog # 10136-044
Supplier:  Enzo Life Sciences
CAS Number:  
Organelle-ID RGB® Reagent IV for Microscopy, Enzo Life Sciences
Catalog # 10136-044
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-53009-C200
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Reagent IV for Microscopy ORGANELLE-ID-RGB®
  • Size:
    200 µL
  • Purity:
    >=93% (HPLC)
  • Cat. No.:
    10136-044
  • Supplier no.:
    ENZ-53009-C200

Specifications

About this item

Mixture of fluorescent dyes for detection of lysosomes, endoplasmic reticulum and nucleic acids

The Organelle-ID RGB® Reagent IV is a mixture of cell-permeable red fluorescent endoplasmic reticulum dye, green fluorescent lysosomal dye and blue fluorescent nucleic acid dye. The staining pattern arising from the combination of these three dyes permits visualization of the target organelles by fluorescence/confocal microscopy. The reagent, supplied as a 500X solution, is sufficient for 1000 microscopy assays. The single-tube format makes this multi-organelle stain reagent easy to use.