Order Entry
Puerto Rico
ContactUsLinkComponent
BioArray™ 5-hmC Methylation Kit, Enzo Life Sciences
BioArray™ 5-hmC Methylation Kit, Enzo Life Sciences
Catalog # 10136-008
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
BioArray™ 5-hmC Methylation Kit, Enzo Life Sciences
Catalog # 10136-008
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-45011-0050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Methylated DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    BioArray™ 5-hmC Methylation Kit
  • Size:
    50 Tests
  • Description:
    BioArray™ 5-hmC Methylation Kit
  • Cat. No.:
    10136-008
  • Supplier no.:
    ENZ-45011-0050

Specifications

About this item

Detect 5-hydroxymethylcytosine using a robust glucosylation method

The BioArray™ 5-hmC methylation kit allows for sequence specific detection of 5-hydroxymethylcytosine within DNA using a simple and efficient glucosylation reaction procedure.

Ordering information: Contains 5-hmC Glucosyltransferase (5-hmC), 10X 5-hmC Reaction buffer, 10X UDPG (uridine diphosphoglucose), MspI restriction enzyme, 5-hmC Control DNA, qPCR Primer 1, qPCR Primer 2 and 5-DNA clean up concentrator kit .