ContactUsLinkComponent
Express DNA Methylation Kit, Shallow Well, Enzo Life Sciences
Express DNA Methylation Kit, Shallow Well, Enzo Life Sciences
  10136-002
 :  
undefined
Express DNA Methylation Kit, Shallow Well, Enzo Life Sciences
  10136-002
 :  ENZ-45003-0002
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Express DNA Methylation Kit, Shallow Well (2x96)
  • Cat. No.:
    10136-002
  • Supplier no.:
    ENZ-45003-0002
Fastest method for complete, high-throughput (96-well) bisulfite conversion of DNA for methylation analysis.
The Express DNA methylation kit (shallow well) features high-throughput (96-well) bisulfite treatment and conversion of DNA for methylation analysis. No preparation is necessary when using the Express Conversion Reagent. Simply add this reagent to a DNA sample and let the reaction proceed to completion. DNA denaturation and bisulfite conversion processes are combined with added heat to facilitate rapid denaturation. Desulphonation and clean-up of the converted DNA is performed using a unique 96-well spin-plate. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
 : Contains Express conversion reagent, Methylation binding buffer, Methylation wash buffer concentrate, L-Desulphonation reagent, Methylation elution buffer, Express binding plate (shallow), Express conversion plates, Collection plates, Elution plates.