DNMT Colorimetric Drug Discovery Kit, Enzo Life Sciences
Catalog # 10135-968
Supplier: Enzo Life Sciences
Specifications
- Description:DNMT colorimetric drug discovery tests
- Cat. No.:10135-968
- No. of reactions:96
- Supplier no.:45017-0001
Specifications
About this item
DNMT colorimetric drug discovery kit is a simple method to detect DNA methylation activity or inhibition.
- Easy colorimetric assay without radioactivity or chromatography
- Measure as little as 0.5 μg of nuclear extract or 0,5 ng of purified enzymes
- Optimised for high specificity to 5-mC without cross-reactivity to unmethylated cytosine
- Reproducible results in less than four hours
Suitable for the quantitative determination of DNMT activity/inhibition in nuclear extracts or purified enzymes.
Ordering information: Includes wash buffer, DNMT assay buffer, adomet, DNMT enzyme control, capture Ab, detection Ab, enhancer solution, developer solution, stop solution, eight-well assay strips with frame, adhesive covering film.