Blood and Tissue DNA Methylation Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
Certificates
About this item
Blood and Tissue DNA Methylation Kit is a simple and reliable DNA bisulfite conversion directly from blood, tissue and cells without the prerequisite of DNA purification.
- Compatible with small sample inputs - as few as 10 cells or 50 pg DNA
- Suited for FFPE and LCM-derived samples with easy to follow instructions
Desulphonation and clean-up of the converted DNA is performed using a low-elution spin column. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Ordering information: Kit contains Proteinase K and storage buffer, Methylation digestion buffer (2X), BT conversion reagent, Methylation dilution buffer, Methylation-solubilisation buffer, Methylation reaction buffer, Methylation binding buffer, Methylation wash buffer concentrate, M-desulphonation buffer, Methylation elution buffer, IC columns and collection tubes.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Methylated DNA
- Sample type:BloodCultured cellsHuman/animal cells and tissues
- Target molecule:DNA
- Description:Blood and Tissue DNA Methylation Kit
Specifications
Frequently Bought Together