ContactUsLinkComponent
Bacto™ Tryptone, Life Technologies
Bacto™ Tryptone, Life Technologies
  90000-286
 :  Avantor
 :  
undefined
Bacto™ Tryptone, Life Technologies
  90000-286
 :  Avantor
 :  
  • Description:
    Tryptone
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. No.:
    90000-286
  • Supplier no.:
    211705
Bacto™ Tryptone is an animal-origin, pancreatic digest of casein.
  • Animal origin
  • Pancreatic digest of casein
  • Included in standard methods and manual applications
  • Provides a range of nutrients
  • Successfully supports the growth of many bacterial species during the production of animal health vaccines
Bacto™ Tryptone is an animal-origin, pancreatic digest of casein. It is included in standard methods and manual applications and is listed in the Reagent section of the United States Pharmacopeia, as meeting the specifications for pancreatic digest of casein, a component in many of the media listed. The nutritional profile of this supplement is unique in that it provides a range of nutrients. Customers have used this supplement in fermentation applications and shown it successfully supports the growth of many bacterial species during the production of animal health vaccines, including Escherichia coli, and Fusobacterium and Agrobacterium species when used together with Bacto™ Yeast Extract.