- Environmentally Preferable:
- Sensitivity:6.03 pg/mL (range 7.81 - 1,000 pg/mL)
- Cat. No.:89141-748
- No. of wells:96 Wells
- Supplier no.:910-001
For the quantitative determination of PGE2 in culture supernatants, serum, saliva, urine, and whole blood from any species.
The PGE2 CLIA kit is a sensitive chemiluminescent competitive enzyme immunoassay kit for the quantitative determination of PGE2 in biological fluids from any species.