TNF Alpha (Mouse) ELISA Kit, Enzo Life Sciences
Catalog # 89141-100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Primary antibody reactivity:Mouse
- Sensitivity:3.9pg/mL (range 31.25 - 2,000pg/mL)
- Cat. No.:89141-100
- Supplier no.:900-047
- No. of tests:96 wells
Specifications
About this item
For the quantitative determination of mouse TNF-α in culture supernatants and serum
The TNF-α (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 5 hours. Absorbance is read at 450nm.
Cited sample type includes tissue and plasma.
Ordering information: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 16, Wash buffer concentrate, Standard, TMB Substrate, Stop solution.