Order Entry
Puerto Rico
ContactUsLinkComponent
Hydrogen Peroxide Colorimetric Detection Kit, Enzo Life Sciences
Hydrogen Peroxide Colorimetric Detection Kit, Enzo Life Sciences
Catalog # 89141-674
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Hydrogen Peroxide Colorimetric Detection Kit, Enzo Life Sciences
Catalog # 89141-674
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-907-015
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Sensitivity:
    51.25 ng/mL (range 106.25 - 3,400 ng/mL)
  • Cat. No.:
    89141-674
  • Supplier no.:
    907-015
  • No. of tests:
    96 wells

Specifications

About this item

For the quantitative determination of hydrogen peroxide in culture supernatants, serum, plasma, and urine from any species

The Hydrogen Peroxide colorimetric detection kit provides a fast and higher throughput 96-well plate based homogeneous assay for the measurement of hydrogen peroxide in a wide variety of samples.

Cited sample type includes tissue.

Ordering information: Contains Half-Area Microtiter plate, Standard, Color reagent.