To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:HighDef® IHC Fluoromount
- Size:25 mL
- Cat. No.:10051-960
- Supplier no.:950-260-0025
Specifications
About this item
Permanent, high stability mounting medium for high definition visualization of IHC.
Liquid. Mounting medium containing <0.1% sodium azide.
HighDef® IHC fluoromount is for mounting tissue specimens stained with fluorescent dyes. Designed for use in fluorescence, but can be used with other applications.