ContactUsLinkComponent
IHC PBS Tablets, Enzo Life Sciences
  10049-368
 :  
IHC PBS Tablets, Enzo Life Sciences
  10049-368
 :  950-240-0100
 :  
  • Description:
    IHC PBS Tablets, 100 pcs
  • Size:
    100 mL
  • Cat. No.:
    10049-368
  • Supplier no.:
    950-240-0100
Solid
100 ml PBS (1X) tablets containing 137 mM sodium chloride, 2.7 mM potassium chloride, and 10 mM phosphate buffer.