Plastic Tube Holder for Two Tubes, Therapak®
Catalog # TPK56590G
Supplier: Therapak, LLC
Specifications
- Description:Tube Holder, PVC, for Two Tubes
- Cat. No.:TPK56590G
- Supplier no.:56590G
Specifications
About this item
This two-tube holder protects fragile specimens or evacuated blood tubes on outbound and inbound shipments
Ideal for inclusion in applications concerning clinical trial specimen tube transport or DUI substance abuse test kits where glass fabricated tubes with gray stopper plugs are used and require protection. Holds tube sizes up to 16 x 100 mm.