Order Entry
United States
ContactUsLinkComponent
Direct cGMP ELISA Kit, Enzo Life Sciences
Direct cGMP ELISA Kit, Enzo Life Sciences
Catalog # 89141-284
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Direct cGMP ELISA Kit, Enzo Life Sciences
Catalog # 89141-284
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-901-014
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type (ELISA with LOV):
    Direct
  • Host:
  • Primary antibody reactivity:
  • Target protein:
    cGMP
  • Environmentally Preferable:
  • Sensitivity:
    (non-acetylated) 0.604 pmol/mL; (acetylated) 0.025 pmol/mL (range (non-acetylated) 0.8 - 500 pmol/mL; (acetylated) 0.08 - 50 pmol/mL)
  • Cat. No.:
    89141-284
  • Supplier no.:
    901-014
  • No. of tests:
    96 wells

Specifications

About this item

Most sensitive (0.025 pmol/mL) colorimetric kit for intracellular cGMP quantification.

For the quantitative determination of cGMP in cell lysates and tissue from any species. Cited sample types include cerebral spinal fluid and microdialysate.

This is a colorimetric competitive immunoassay kit for the quantitation of intracellular cGMP in cells or tissues lysed in 0.1 M HCl, which is used to stop endogenous phosphodiesterase activity and stabilize the released cyclic GMP. The 0.1M HCl treated samples are then analyzed directly in a microtiter plate without extraction, drying and reconstitution. Sensitivity is increased by acetylation (reagents included). Absorbance is read at 405 nm.The ready-to-use liquid color-coded reagents provided reduce error.

Ordering information: Contains GxR Microtiter plate, Conjugate, Antibody, 0.1M HCl, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Acetic anhydride, Triethylamine. Alternative size available: 89141-068 (1x96 wells).