Shipper, EAS Ambient, Therapak®
10029-408
- Description:Teartech® EAS ambient specimen shipper
- Cat. No.:10029-408
- Supplier no.:56879
For use in both regional and national Veterinary testing laboratories and Veterinary hospitals, the EAS specimen shipper features a bulk-pack case of shipping supplies to transport ambient samples
The shipper includes: 50 TearTech® easy to assemble boxes, 100 zip-style biohazard transport bags (two pocket bag for sample and requisition form), 100 Aqui-Pak™ segmented absorbent pouches (each Aqui-Pak™ holds up to 6 standard blood or urine tubes), 50 biohazard labels, 50 Exempt Animal Specimen labels, and cushioning material where applicable.
Each TearTech® EAS shipper box holds up to four specimen bags with patient samples.