Shipper, EHS Frozen, Therapak®
Catalog # 10029-404
Supplier: Therapak, LLC
Specifications
- Description:EHS frozen shipping system
- Cat. No.:10029-404
- Supplier no.:56877
Specifications
About this item
EHS shipping systems for frozen specimens feature Therapak®'s standard small insulated cooler (ID: 8 x 6 x 8”) and box (OD: 11 x 9 x 11”) with a Class 9 Dry Ice label, making it ideal for overnight shipments.
Includes specimen bag, absorbent material, Exempt Human Specimen label, biohazard symbol label, and cushioning material. Add sample tubes and dry ice to ship.