Order Entry
Puerto Rico
ContactUsLinkComponent
Drawer Organizer Utility, 4-Compartment, Polystyrene, TrippNT
Drawer Organizer Utility, 4-Compartment, Polystyrene, TrippNT
Catalog # 10012-704
Supplier:  TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Drawer Organizer Utility, 4-Compartment, Polystyrene, TrippNT
Catalog # 10012-704
Supplier:  TrippNT
Supplier Number:  50031
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    4-Compartment Utility Drawer Organizer
  • Cat. No.:
    10012-704
  • Ext. dimensions:
    48.6W×6H×44.5D cm (19 1/8×2 3/8×17 1/2")
  • Weight:
    0.45 kg (1 lbs.)
  • Supplier no.:
    50031

Specifications

About this item

Styrene drawer organizer for small items or drawers. Comes with 4 pockets.

  • Chemically resistant
  • 2 Sizes available
  • In stock and ready to ship
  • American made by TrippNT
  • Total replacement Lifetime Guarantee

Can be used for storage or as a tray. Built in lip for easy carrying.

White color. Styrene material.