Samco™ Bio-Tite™ Specimen Container for Operating Room, Thermo Scientific
Catalog # 14231-916
Supplier: Samco
Specifications
- Cap Size:53 mm
- Sterility:Sterile
- Capacity:120 ml
- Cat. No.:14231-916
- Packaging:Individually Wrapped (Poly Bag)
- Cap color:Blue
- Supplier no.:07 0006
Specifications
About this item
Samco™ Bio-Tite™ Specimen containers for the operating room.
- Supplied sterile
- Shatter-resistant containers are manufactured from virgin polypropylene
- Leak-resistant seal prevents bio-hazardous spills
Container is ideal for collecting biopsy specimens and transporting them into a sterile environment. It is tested at varying pressures, making it ideal for use in various transport systems.
Certifications: FDA Class 1 Exempt Medical Device.
Ordering information: Ambient storage and shipping recommended for containers.