Order Entry
Puerto Rico
ContactUsLinkComponent
Acid Capacity Cell Test To pH 4.3 (Total Alkalinity), Spectroquant®, MilliporeSigma
Acid Capacity Cell Test To pH 4.3 (Total Alkalinity), Spectroquant®, MilliporeSigma
Catalog # EM1.01758.0001
Supplier:  MilliporeSigma
CAS Number:  
Acid Capacity Cell Test To pH 4.3 (Total Alkalinity), Spectroquant®, MilliporeSigma
Catalog # EM1.01758.0001
Supplier:  MilliporeSigma
Supplier Number:  1.01758.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Chemical method:
    Indicator
  • Method:
    Photometric 0.40 - 8.00 mmol/l 20 - 400 mg/l CaCO3
  • Storage Temperature:
    Store at +15°C to +25°C.
  • Cat. No.:
    EM1.01758.0001
  • Range:
    0.40 - 8.00 mmoL/L 20 - 400 mg/L CaCO₃
  • Supplier no.:
    1.01758.0001
  • No. of tests:
    120
  • For unit:
    Prove, Nova 30/60, Multy, Move

Specifications

About this item