ContactUsLinkComponent
Shipper, Category B Frozen, Large Canister, Therapak®
Shipper, Category B Frozen, Large Canister, Therapak®
Catalog # 10029-302
Supplier:  Therapak, LLC
undefined
Shipper, Category B Frozen, Large Canister, Therapak®
Catalog # 10029-302
Supplier:  Therapak, LLC
Supplier Number:  37817
Specifications
  • Description:
    Cat B frozen, Large canister
  • Cat. No.:
    10029-302
  • Supplier no.:
    37817
Specifications
About this item
Frozen specimen shipping system.
  • Holds up to eighteen tubes for overnight shipments
Includes transport box (OD: 15x13x12"), medium cooler (ID: 12x10x9"), large 95kPa canister, three Aqui-Pak™ segmented absorbent pouches, cushioning material, list of contents card, dry ice label, and required labels for IATA PI650 marking and compliance.