Shipper, Category B Ambient, 95 kPa Bag, Zorb Sheet
Catalog # 10029-280
Supplier: Therapak, LLC
Specifications
- Description:Cat B ambient, 95 kPa bag, zorb sheet
- Cat. No.:10029-280
- Supplier no.:37812
Specifications
About this item
Ideal for transporting multiple or single non-fragile items such as specimen containers or 20 to 50 ml sample vials.
Delivery: The ambient transport system for blood and urine tube transport includes: 95 kPa bag, absorbent sheet, cushioning material, list of contents card, biohazard symbol label, biological substance category B label, and medium transport box (int.Ø: 7×4×4"; ext.Ø: 8×4.25×4.25")