Order Entry
Puerto Rico
ContactUsLinkComponent
Fluoride Test, MQuant®, MilliporeSigma
Fluoride Test, MQuant®, MilliporeSigma
Catalog # EM1.18771.0001
Supplier:  MilliporeSigma
CAS Number:  
Fluoride Test, MQuant®, MilliporeSigma
Catalog # EM1.18771.0001
Supplier:  MilliporeSigma
Supplier Number:  1.18771.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Chemical method:
    Alizarin complexone
  • Method:
    Colorimetric with color card 0 - 0.15 - 0.3 - 0.5 - 0.8 mg/l F
  • Storage Temperature:
    Store at +15°C to +25°C.
  • Cat. No.:
    EM1.18771.0001
  • Range:
    0 - 0.15 - 0.3 - 0.5 - 0.8 mg/L F
  • Supplier no.:
    1.18771.0001
  • No. of tests:
    100

Specifications

About this item

Quick tests, visual colour comparison

  • Results in approximately 5 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.