Order Entry
Puerto Rico
ContactUsLinkComponent
Nitrite Test, MColortest™, MilliporeSigma
Nitrite Test, MColortest™, MilliporeSigma
Catalog # EM1.14424.0001
Supplier:  MilliporeSigma
CAS Number:  
Nitrite Test, MColortest™, MilliporeSigma
Catalog # EM1.14424.0001
Supplier:  MilliporeSigma
Supplier Number:  1.14424.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Chemical method:
    Griess' reaction
  • Method:
    Colorimetric with color card and comparator block with short test tubes 0.1-0.2-0.3-0.4-0.6-0.8-1.0-1.3-2.0 mg/l NO₂⁻ 0.03-0.06-0.09-0.12-0.18-0.24-0.3-0.4-0.6 mg/l NO₂-N
  • Storage Temperature:
    Store at +15°C to +25°C.
  • Cat. No.:
    EM1.14424.0001
  • Range:
    0.1-0.2-0.3-0.4-0.6-0.8-1.0-1.3-2.0 mg/L NO₂⁻ 0.03-0.06-0.09-0.12-0.18-0.24-0.3-0.4-0.6 mg/L NO₂-N
  • Supplier no.:
    1.14424.0001
  • No. of tests:
    400

Specifications

About this item

Nitrite is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications

Nitrites must be closely monitored in drinking and ground water as well as in process, waste, or aquarium water. In farm produce and fruit juices, high nitrate concentrations affect quality and keeping properties and can constitute serious health risks. Nitrites are also used in additives for food products and serve as corrosion inhibitors in engine cooling systems.