ContactUsLinkComponent
Spectroquant® AOX Test Kits, MilliporeSigma
Spectroquant® AOX Test Kits, MilliporeSigma
  EM1.00677.0001
 :  
undefined
Spectroquant® AOX Test Kits, MilliporeSigma
  EM1.00677.0001
 :  1.00677.0001
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Chemical method:
    Iron(III) thiocyanate
  • Description:
    AOX sample preparation set
  • Method:
    Photometric 0.05 - 2.50 mg/l
  • Storage Temperature:
    Store at 15...25 °C
  • Cat. No.:
    EM1.00677.0001
  • Range:
  • Supplier no.:
    1.00677.0001
  • No. of tests:
    25
  • For unit:
    Prove, Nova 30/60, Multy, Move
Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.