ContactUsLinkComponent
Phosphate Cell Test, Spectroquant®, MilliporeSigma
Phosphate Cell Test, Spectroquant®, MilliporeSigma
Catalog # EM1.14546.0001
Supplier:  MilliporeSigma
CAS Number:  
Phosphate Cell Test, Spectroquant®, MilliporeSigma
Catalog # EM1.14546.0001
Supplier:  MilliporeSigma
Supplier Number:  1.14546.0001
CAS Number:  
Specifications
  • Chemical method:
    Vanadate-molybdate
  • Pack type:
    EPS-Moulding
  • Size:
    1 pc
  • Cat. No.:
    EM1.14546.0001
  • Range:
    0.5 - 25.0 mg/L PO₄-P, 1.5 - 76.7 mg/L PO₄,
    1.1 - 57.3 mg/L P₂O₅
  • Supplier no.:
    1.14546.0001
  • No. of tests:
    25
  • For unit:
    Prove, Nova 30/60, Multy, Move
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).