Order Entry
Puerto Rico
ContactUsLinkComponent
MColortest™ Oxygen Test, MilliporeSigma
MColortest™ Oxygen Test, MilliporeSigma
Catalog # EM1.11152.0001
Supplier:  MilliporeSigma
CAS Number:  
MColortest™ Oxygen Test, MilliporeSigma
Catalog # EM1.11152.0001
Supplier:  MilliporeSigma
Supplier Number:  1.11152.0001
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Chemical method:
    Winkler
  • Description:
    Oxygen Test Refill Pack
  • Method:
    Titrimetric, acc. to Winkler, with titration pipette
  • Storage Temperature:
    Store at +15°C to +25°C.
  • Cat. No.:
    EM1.11152.0001
  • Supplier no.:
    1.11152.0001
  • No. of tests:
    100

Specifications

About this item

The sustainability of biological processes in natural waters is essentially depended on a sufficient oxygen content

When the reagents of the MColortestTM test are used up, the kit can be refilled by ordering the appropriate refill pack. They are available for a range of MColortestTM test kits, making it a particularly economical system.

The oxygen present in water is also consumed by chemical processes and by microorganisms.