ContactUsLinkComponent
Mouse Trapeze, Bio-Serv
  89412-130
 :  Bio-Serv
undefined
Mouse Trapeze, Bio-Serv
  89412-130
 :  Bio-Serv
 :  K3614
  • Width:
    4.92 cm (1¹⁵⁄₁₆")
  • Description:
    Mouse Trapeze
  • Length:
    11.1 cm (4⅜")
  • Environmentally Preferable:
  • Cat. No.:
    89412-130
  • Supplier no.:
    K3614
Promotes climbing and 3-Dimensional use of cage
Made of stainless steel. Allows safe retreat in flooded cages.