Mouse Diet, High Fat Calories, Bio-Serv
Supplier: Bio-Serv
Certificates
About this item
Nutritionally complete, purified diet formula successfully used in diabetes and obesity studies
- 60% kcals from fat. For induction and/or maintenance of obese models
- Nutritionally assayed
- Shelf Life: 6 months, refrigerated
Ingredients: Lard, Sucrose, Casein, Maltodextrin, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride.
Caloric value: 5.51 kcal/g for paste, 5.49 kcal/g for pellets
Nutritional ash content: 3.5%
Nutritional carbohydrate content: 36.2% for paste, 35.7% for pellets
Nutritional fat content: 36.0%
Nutritional fibre content:
Nutritional moisture content: <10%
Nutritional protein content: 20.5%