Specifications
- Ø:2.54 cm (1")
- Description:SCIDS MD's, Bacon Flavor, Sterile
- Height:0.64 cm (¼")
- Size:5 g tablet
- Cat. No.:89067-458
- Packaging:100/Bag
- Supplier no.:S0443
Specifications
About this item
An effective method to reduce the hazards of respiratory problems in immunocompromised mice
- Bacon-flavored tablets are highly palatable, nutritionally complete, and grain-based
- More convenient and minimizes risk of dehydration compared to using medicated water
- Direct compression process eliminates drug degradation
- Sterilized by gamma irradiation
- Nutritionally assayed
- Shelf Life: 12 months, cool dry storage.
Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin, Sulfamethoxazole, Trimethoprim.
Each bacon-flavored, scored tablet contains 60 mg Sulfamethoxazole and 12 mg Trimethoprim. Mouse average dose: One 5 g tab/20 g of body weight. To administer, place on cage floor for easy access.
Caloric value: 3.39 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 4.3%
Nutritional fibre content: 4.0%
Nutritional moisture content: <10%
Nutritional protein content: 21.3%