Specifications
- Ø:2.54 cm (1")
- Description:Rat Helicobacter 4 Drug Combo Tablet
- Height:0.64 cm (¼")
- Size:5 g tablet
- Cat. No.:89067-454
- Packaging:100/Bag
- Supplier no.:F05648
Specifications
About this item
Most effective oral treatment against a broad spectrum of Helicobacter strains
- Highly palatable, bacon-flavored, nutritionally complete diet
- Used to treat breeders or quarantined rodents
- Alternative method of treatment to cesarean rederivation
- Comprehensive husbandry plan must be employed for success; contact Bio-Serv for details
- Nutritionally assayed
- Shelf Life: 6 months, cool dry storage
Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin, Omeprazole, Metronidazole, Amoxicillin, Clarithromycin.
Each 5 g Rat Helicobacter MD tablet contains 10.00 mg of Amoxicillin, 1.70 mg of Clarithromycin, 3.30 mg of Metronidazole and 0.07 mg of Omeprazole. Rat average dose: One 5 g tablet/100 g of body weight. Available in pellets. Sterilization available by gamma irradiation.
Caloric value: 3.39 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 4.2%
Nutritional fibre content: 3.9%
Nutritional moisture content: <10%
Nutritional protein content: 21.2%