Specifications
- Ø:2.54 cm (1")
- Description:Mouse Helicobacter 4 Drug Combo Tablet
- Height:0.64 cm (¼")
- Size:5 g tablet
- Cat. No.:89067-452
- Packaging:100/Bag
- Supplier no.:S05723
Specifications
About this item
The most effective oral treatment against a broad spectrum of Helicobacter strains
- Highly palatable, bacon-flavored, nutritionally complete diet. Used to treat breeders or quarantined mice
- Alternative method of treatment to cesarean rederivation
- Comprehesive husbandry plan must be employed for success; contact Bio-Serv for details
- Nutritionally assayed
- Shelf Life: 12 months, cool dry storage
Ingredients: Ground Corn, Dehulled Soybean Meal, Dried Beet Pulp, Fish Meal, Ground Oats, Brewers Dried Yeast, Cane Molasses, Dehydrated Alfalfa Meal, Dried Whey, Wheat Germ, Porcine Animal Fat (preserved with BHA), Porcine Meat Meal, Wheat Middlings, Mineral Mix, Vitamin Mix, Bacon Flavor, DL-Methionine, Choline Chloride, Omeprazole, Metronidazole, Amoxicillin, Clarithromycin.
Each 5 g Mouse Helicobacter MD contains 3.00 mg of Amoxicillin, 0.50 mg of Clarithromycin, 1.00 mg of Metronidazole and 0.02 mg of Omeprazole. Mouse average dose: One 5 g tablet/50 g of body weight.
Caloric value: 3.39 kcal/g
Nutritional ash content: 7.8%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 4.3%
Nutritional fibre content: 4.0%
Nutritional moisture content: <10%
Nutritional protein content: 21.3%